Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277399.1 | NP_001264328.1 | 1 - 2 | 144 | 59 | |
GenBank mRNA | BU472291.1 | - | 1 - 2 | 140 | 59 | |
BX934448.1 | - | 1 - 2 | 110 | 59 | ||
CR353120.1 | - | 1 - 2 | 117 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |