Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277399.1 | NP_001264328.1 | 3 - 4 | 305 | 95 | |
GenBank mRNA | BX934448.1 | - | 3 - 4 | 271 | 95 | |
CR353120.1 | - | 3 - 4 | 278 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |