Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277399.1 | NP_001264328.1 | 5 - 6 | 470 | 116 | |
GenBank mRNA | BX934448.1 | - | 5 - 6 | 436 | 116 | |
CR353120.1 | - | 5 - 6 | 443 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |