Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419331.6 | XP_419331.1 | 5 - 6 | 586 | 82 | |
GenBank mRNA | BX931436.2 | - | 5 - 6 | 567 | 82 | |
BX932844.2 | - | 5 - 6 | 568 | 82 | ||
BX933982.1 | - | 5 - 6 | 577 | 82 | ||
BX934618.1 | - | 5 - 6 | 580 | 82 | ||
BX935753.1 | - | 5 - 6 | 574 | 82 |
Gene Symbol: CTAGE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025148519.1 | XP_025004287.1 | 2 - 3 | 139 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |