Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBNDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001232717.5 | XP_001232718.2 | 3 - 4 | 393 | 77 | |
XM_003641947.4 | XP_003641995.1 | 4 - 5 | 483 | 77 | ||
XM_004944325.3 | XP_004944382.1 | 4 - 5 | 933 | 77 | ||
XM_004944326.3 | XP_004944383.1 | 3 - 4 | 861 | 77 | ||
XM_004944327.3 | XP_004944384.1 | 3 - 4 | 843 | 77 | ||
XM_004944328.3 | XP_004944385.1 | 4 - 5 | 480 | 77 | ||
XM_015292686.2 | XP_015148172.1 | 4 - 5 | 951 | 77 | ||
XM_015292687.2 | XP_015148173.1 | 3 - 4 | 858 | 77 | ||
XM_015292688.2 | XP_015148174.1 | 4 - 5 | 906 | 77 | ||
XM_015292689.2 | XP_015148175.1 | 4 - 5 | 906 | 77 | ||
XM_025154249.1 | XP_025010017.1 | 4 - 5 | 991 | 77 | ||
XM_025154250.1 | XP_025010018.1 | 3 - 4 | 813 | 77 | ||
GenBank mRNA | BX935557.1 | - | 4 - 5 | 552 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |