Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CETN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277920.1 | NP_001264849.1 | 3 - 4 | 345 | 117 | |
GenBank mRNA | BX930994.2 | - | 3 - 4 | 345 | 117 | |
BX935748.1 | - | 3 - 4 | 347 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |