Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004945796.3 | XP_004945853.2 | 5 - 6 | 808 | 71 | |
XM_015279690.2 | XP_015135176.2 | 5 - 6 | 960 | 71 | ||
XM_025141524.1 | XP_024997292.1 | 5 - 6 | 965 | 71 | ||
XM_415524.6 | XP_415524.4 | 5 - 6 | 813 | 71 | ||
GenBank mRNA | CR353484.1 | - | 5 - 6 | 823 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |