Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277728.2 | XP_015133214.1 | 1 - 2 | 504 | 71 | |
GenBank mRNA | CR354088.1 | - | 1 - 2 | 101 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |