Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLPPR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015290742.2 | XP_015146228.1 | 4 - 5 | 615 | 78 | |
XM_015290743.2 | XP_015146229.1 | 1 - 2 | 343 | 78 | ||
XM_015290744.2 | XP_015146230.1 | 2 - 3 | 1046 | 78 | ||
XM_025152941.1 | XP_025008709.1 | 1 - 2 | 343 | 78 | ||
XM_422320.5 | XP_422320.2 | 1 - 2 | 343 | 78 | ||
GenBank mRNA | BX930216.2 | - | 1 - 2 | 267 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |