Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277753.1 | NP_001264682.1 | 3 - 4 | 588 | 113 | |
XM_025149971.1 | XP_025005739.1 | 3 - 4 | 594 | 113 | ||
GenBank mRNA | BX932128.2 | - | 3 - 4 | 421 | 113 | |
BX933163.2 | - | 3 - 4 | 588 | 113 | ||
CN235120.1 | - | 3 - 4 | 588 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |