Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015275978.2 | XP_015131464.1 | 4 - 5 | 1319 | 80 | |
XM_015275981.2 | XP_015131467.1 | 4 - 5 | 1371 | 80 | ||
XM_015275985.2 | XP_015131471.1 | 4 - 5 | 1402 | 80 | ||
XM_417078.5 | XP_417078.1 | 4 - 5 | 1403 | 80 | ||
GenBank mRNA | BX933025.2 | - | 2 - 3 | 556 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |