Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPH14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415246.6 | XP_415246.1 | 5 - 6 | 937 | 79 | |
GenBank mRNA | BX934413.2 | - | 5 - 6 | 919 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |