Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278123.1 | NP_001265052.1 | 2 - 3 | 374 | 103 | |
XM_015298002.2 | XP_015153488.1 | 2 - 3 | 387 | 103 | ||
GenBank mRNA | BX935609.2 | - | 2 - 3 | 375 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |