Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278123.1 | NP_001265052.1 | 4 - 5 | 699 | 71 | |
XM_015298002.2 | XP_015153488.1 | 4 - 5 | 712 | 71 | ||
GenBank mRNA | BX935609.2 | - | 4 - 5 | 700 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |