Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277823.1 | NP_001264752.1 | 3 - 4 | 400 | 98 | |
NM_001277824.1 | NP_001264753.1 | 3 - 4 | 320 | 98 | ||
GenBank mRNA | AM067556.1 | - | 3 - 4 | 320 | 98 | |
BX932596.2 | - | 3 - 4 | 400 | 98 | ||
BX934467.2 | - | 3 - 4 | 293 | 98 | ||
BX936078.1 | - | 3 - 4 | 312 | 98 | ||
BX936226.2 | - | 3 - 4 | 321 | 98 | ||
CN219349.1 | - | 3 - 4 | 308 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |