Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419831.6 | XP_419831.2 | 1 - 2 | 85 | 89 | |
GenBank mRNA | BX930133.1 | - | 1 - 2 | 50 | 89 | |
CR354351.1 | - | 1 - 2 | 68 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |