Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX929565.1 | - | 2 - 3 | 245 | 104 | |
CR385154.1 | - | 1 - 2 | 75 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |