Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004949515.3 | XP_004949572.1 | 12 - 13 | 1268 | 143 | |
XM_004949517.3 | XP_004949574.1 | 11 - 12 | 1147 | 143 | ||
XM_015280701.1 | XP_015136187.1 | 12 - 13 | 1320 | 143 | ||
XM_015280702.1 | XP_015136188.1 | 13 - 14 | 1469 | 143 | ||
XM_015280703.2 | XP_015136189.1 | 12 - 13 | 1266 | 143 | ||
XM_015280704.2 | XP_015136190.1 | 12 - 13 | 1469 | 143 | ||
XM_025144918.1 | XP_025000686.1 | 11 - 12 | 1199 | 143 | ||
XM_424912.6 | XP_424912.3 | 12 - 13 | 1230 | 143 | ||
GenBank mRNA | CR385816.1 | - | 5 - 6 | 418 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |