Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414186.6 | XP_414186.5 | 1 - 2 | 1207 | 65 | |
GenBank mRNA | AB028627.1 | - | 1 - 2 | 750 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |