Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257283.3 | NP_001244212.1 | 2 - 3 | 913 | 62 | |
NM_001319309.2 | NP_001306238.1 | 2 - 3 | 913 | 62 | ||
XM_025143394.1 | XP_024999162.1 | 3 - 4 | 4391 | 62 | ||
GenBank mRNA | AF120210.1 | - | 2 - 3 | 511 | 62 | |
CR390222.1 | - | 2 - 3 | 473 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |