Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271931.3 | NP_001258860.2 | 6 - 7 | 2219 | 80 | |
GenBank mRNA | AY283064.1 | - | 3 - 4 | 1263 | 80 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Gallus gallus Probe spans exons |