Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015276388.2 | XP_015131874.1 | 4 | 4430 | 101 | |
GenBank mRNA | BX929304.1 | - | 1 | 1446 | 101 | |
BX929306.1 | - | 1 | 1446 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Both primers and probe map within a single exon |