Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLT8D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277383.2 | NP_001264312.2 | 7 - 8 | 894 | 68 | |
GenBank mRNA | BX929886.1 | - | 7 - 8 | 894 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |