Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERICH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278029.1 | NP_001264958.1 | 4 - 5 | 804 | 86 | |
XM_015284836.2 | XP_015140322.1 | 4 - 5 | 777 | 86 | ||
XM_015284837.2 | XP_015140323.2 | 4 - 5 | 821 | 86 | ||
GenBank mRNA | BX930034.1 | - | 4 - 5 | 767 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |