Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277747.2 | XP_015133233.1 | 5 - 6 | 755 | 138 | |
XM_015277748.2 | XP_015133234.1 | 6 - 7 | 789 | 138 | ||
XM_015277749.2 | XP_015133235.1 | 6 - 7 | 672 | 138 | ||
XM_015277750.2 | XP_015133236.1 | 7 - 8 | 1095 | 138 | ||
XM_015277751.2 | XP_015133237.1 | 6 - 7 | 1225 | 138 | ||
XM_015277752.2 | XP_015133238.1 | 5 - 6 | 736 | 138 | ||
XM_015277753.2 | XP_015133239.1 | 5 - 6 | 624 | 138 | ||
XM_015277754.2 | XP_015133240.1 | 5 - 6 | 736 | 138 | ||
GenBank mRNA | CR389339.1 | - | 2 - 3 | 263 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |