Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277747.2 | XP_015133233.1 | 7 - 8 | 1134 | 99 | |
XM_015277748.2 | XP_015133234.1 | 8 - 9 | 1168 | 99 | ||
XM_015277749.2 | XP_015133235.1 | 8 - 9 | 1051 | 99 | ||
XM_015277750.2 | XP_015133236.1 | 9 - 10 | 1474 | 99 | ||
XM_015277751.2 | XP_015133237.1 | 8 - 9 | 1604 | 99 | ||
XM_015277752.2 | XP_015133238.1 | 7 - 8 | 1115 | 99 | ||
XM_015277753.2 | XP_015133239.1 | 7 - 8 | 1003 | 99 | ||
XM_015277754.2 | XP_015133240.1 | 7 - 8 | 1115 | 99 | ||
GenBank mRNA | CR389339.1 | - | 4 - 5 | 642 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |