Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JADE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015277747.2 | XP_015133233.1 | 8 - 9 | 1250 | 88 | |
XM_015277748.2 | XP_015133234.1 | 9 - 10 | 1284 | 88 | ||
XM_015277749.2 | XP_015133235.1 | 9 - 10 | 1167 | 88 | ||
XM_015277750.2 | XP_015133236.1 | 10 - 11 | 1590 | 88 | ||
XM_015277751.2 | XP_015133237.1 | 9 - 10 | 1720 | 88 | ||
XM_015277752.2 | XP_015133238.1 | 8 - 9 | 1231 | 88 | ||
XM_015277753.2 | XP_015133239.1 | 8 - 9 | 1119 | 88 | ||
XM_015277754.2 | XP_015133240.1 | 8 - 9 | 1231 | 88 | ||
GenBank mRNA | CR389339.1 | - | 5 - 6 | 758 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |