Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015279008.2 | XP_015134494.1 | 2 - 3 | 437 | 65 | |
XM_015279009.2 | XP_015134495.1 | 2 - 3 | 437 | 65 | ||
XM_025154079.1 | XP_025009847.1 | 2 - 3 | 437 | 65 | ||
GenBank mRNA | CR389722.1 | - | 2 - 3 | 185 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |