Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRPS1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_420181.6 | XP_420181.1 | 1 - 2 | 286 | 76 | |
GenBank mRNA | CR407240.1 | - | 1 - 2 | 280 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |