Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004944268.3 | XP_004944325.1 | 4 - 5 | 126 | ||
XM_004944269.3 | XP_004944326.1 | 5 - 6 | 126 | |||
XM_004944270.3 | XP_004944327.1 | 4 - 5 | 126 | |||
XM_015292625.2 | XP_015148111.1 | 5 - 6 | 126 | |||
XM_015292626.2 | XP_015148112.1 | 3 - 4 | 126 | |||
XM_025154330.1 | XP_025010098.1 | 4 - 5 | 126 | |||
XM_025154331.1 | XP_025010099.1 | 5 - 6 | 126 | |||
XM_025154332.1 | XP_025010100.1 | 5 - 6 | 126 | |||
XM_025154333.1 | XP_025010101.1 | 4 - 5 | 126 | |||
XM_414178.6 | XP_414178.3 | 4 - 5 | 126 | |||
GenBank mRNA | CR406953.1 | - | 1 - 2 | 126 | ||
CR523083.1 | - | 2 - 3 | 705 | 126 | ||
CR523768.1 | - | 1 - 2 | 256 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |