Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199476.1 | NP_001186405.1 | 3 - 4 | 426 | 93 | |
GenBank mRNA | BX935863.1 | - | 3 - 4 | 409 | 93 | |
CR523331.1 | - | 3 - 4 | 426 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |