Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PVALB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | M65068.1 | - | 1 | 62 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Amplicon spans exons and probe does not span exons |