Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMP2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277356.1 | NP_001264285.1 | 1 - 2 | 96 | 60 | |
XM_025147657.1 | XP_025003425.1 | 1 - 2 | 415 | 60 | ||
GenBank mRNA | BX930706.1 | - | 1 - 2 | 93 | 60 | |
BX931538.1 | - | 1 - 2 | 96 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |