Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414494.6 | XP_414494.4 | 1 - 2 | 242 | 114 | |
GenBank mRNA | BX930980.1 | - | 1 - 2 | 226 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |