Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_414494.6 | XP_414494.4 | 2 - 3 | 291 | 99 | |
GenBank mRNA | BX930980.1 | - | 2 - 3 | 275 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |