Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10H15ORF48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX931191.1 | - | 3 - 4 | 338 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |