Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015284385.2 | XP_015139871.1 | 6 - 7 | 3257 | 79 | |
GenBank mRNA | BX931270.1 | - | 5 - 6 | 849 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |