Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419775.6 | XP_419775.1 | 2 - 3 | 301 | 125 | |
GenBank mRNA | BX932116.1 | - | 2 - 3 | 300 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |