Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001354731.2 | NP_001341660.1 | 3 - 4 | 580 | 80 | |
GenBank mRNA | BX931478.1 | - | 4 - 5 | 701 | 80 | |
BX932994.1 | - | 2 - 3 | 337 | 80 | ||
BX950277.1 | - | 2 - 3 | 479 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |