Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004934559.3 | XP_004934616.1 | 2 - 3 | 350 | 132 | |
XM_025145360.1 | XP_025001128.1 | 2 - 3 | 350 | 132 | ||
XM_416733.6 | XP_416733.4 | 2 - 3 | 349 | 132 | ||
GenBank mRNA | BX933172.1 | - | 2 - 3 | 315 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |