Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102328.2 | 198 | 100 | |||
GenBank mRNA | BX933338.1 | - | 199 | 112 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Gallus gallus |