Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277337.1 | NP_001264266.1 | 3 - 4 | 474 | 70 | |
XM_015278680.2 | XP_015134166.1 | 3 - 4 | 333 | 70 | ||
GenBank mRNA | BX934609.1 | - | 3 - 4 | 352 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |