Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_025146874.1 | XP_025002642.1 | 2 - 3 | 650 | 60 | |
XM_417174.6 | XP_417174.1 | 2 - 3 | 418 | 60 | ||
GenBank mRNA | BX935584.1 | - | 2 - 3 | 435 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |