Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277522.1 | NP_001264451.1 | 4 - 5 | 568 | 112 | |
NM_001277526.1 | NP_001264455.1 | 4 - 5 | 509 | 112 | ||
NM_001277528.1 | NP_001264457.1 | 3 - 4 | 360 | 112 | ||
XM_015291998.2 | XP_015147484.1 | 4 - 5 | 478 | 112 | ||
XM_025153839.1 | XP_025009607.1 | 4 - 5 | 395 | 112 | ||
XM_025153840.1 | XP_025009608.1 | 3 - 4 | 303 | 112 | ||
GenBank mRNA | BU109075.1 | - | 4 - 5 | 521 | 112 | |
BU137192.1 | - | 3 - 4 | 310 | 112 | ||
BX935855.1 | - | 4 - 5 | 443 | 112 | ||
CF251301.1 | - | 3 - 4 | 362 | 112 | ||
CN237237.1 | - | 4 - 5 | 568 | 112 | ||
DR411810.1 | - | 4 - 5 | 507 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |