Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPA33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_416656.6 | XP_416656.1 | 6 - 7 | 894 | 63 | |
GenBank mRNA | BX934867.1 | - | 6 - 7 | 819 | 63 | |
BX936257.1 | - | 6 - 7 | 819 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |