Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199598.1 | NP_001186527.1 | 3 - 4 | 520 | 64 | |
GenBank mRNA | BX932258.2 | - | 3 - 4 | 235 | 64 | |
BX950714.1 | - | 3 - 4 | 520 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |