Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLA2G2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277914.1 | NP_001264843.1 | 1 - 2 | 72 | 88 | |
GenBank mRNA | BU270619.1 | - | 1 - 2 | 52 | 88 | |
BX950757.1 | - | 1 - 2 | 72 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |