Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRNKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_419315.6 | XP_419315.2 | 1 - 2 | 86 | 54 | |
GenBank mRNA | CR352411.1 | - | 1 - 2 | 63 | 54 | |
CR388981.1 | - | 1 - 2 | 56 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |