Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277618.3 | NP_001264547.1 | 2 - 3 | 265 | 67 | |
GenBank mRNA | CR352495.1 | - | 2 - 3 | 268 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |