Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_415846.6 | XP_415846.2 | 4 - 5 | 670 | 142 | |
GenBank mRNA | CR352613.1 | - | 4 - 5 | 508 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |